Learn More
Sino Biological Human IFNG Protein 1MG (Sino Biological)
Supplier:  Sino Biological 14074MM01100
Encompass_Preferred
A DNA sequence encoding the human IFNG (NP_000610.2) (Gln24-Gln166) was expressed.
Catalog No. 50-237-4288