Learn More
PBL Assay Science DIY (Do-It-Yourself) Human Interferon Lambda 1/2/3 (IFNL1/IFNL2/IFNL3) / Interleukin-29/28A/B (IL-29/IL-28A/IL-28B) ELISA, Tissue Culture Media (TCM) [Materials for 15x96 well plate]
Supplier:  PBL Assay Science 618401
Encompass
The Do-It-Yourself (DIY) immunoassay Human Interferon Lambda 1/2/3 (Interleukin-29/28A/28B) (IL-29/28A/28B) ELISA kit from PBL Assay Science (Catalog #61840-1) for research use only (RUO) and packaged with materials for 15x96-well plates. This ELISA is Tissue Culture Medium (TCM) compatible and has an assay range of 62.5-4000 pg/ml. This kit contains the basic components necessary for the development of sandwich ELISAs to measure natural and recombinant Human IFN-Lambda 1/2/3 (IL29/IL28A/IL28B, respectively). This kit is most reactive with Lambda 3 and Lambda 1.
Catalog No. 50-153-8080