Learn More
Sigma Aldrich Fine Chemicals Biosciences Human IfNL1 Interferon l
Catalog No. 502904884
Encompass
The Human IL-29 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-29 in serum plasma cell culture supernatants and urine.
Catalog No. 50-290-4884 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. RAB02931KT
The Human IL-29 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-29 in serum plasma cell culture supernatants and urine.