Learn More
Sino Biological Anti-IL-29/IFN-lambda 1 Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 12339MM03H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-29 (rh IL-29; Catalog#12339-H08H; NP_742152.1; Met1-Thr200). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-2518