Learn More
Sino Biological Anti-IL-29/IFN-lambda 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12339T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL-29 (rh IL-29; Catalog#12339-H08H; NP_742152.1; Met1-Thr200). IL-29 specific IgG was purified by Human IL-29 affinity chromatography.
Catalog No. 50-240-7653