Learn More
Sino Biological Anti-IFT20 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 108067T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human IFT20/intraflagellar transport 20, and purified by antigen affinity chromatography.
Catalog No. 50-240-8211