Learn More
Bioss Human Ig lambda light chain Re
SDP
Catalog No. 5000066690
Encompass_Preferred
Human Ig lambda light chain Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IHC-P IHC-F IF(ICC) IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-66690 Supplier Bioss Supplier No. BSM61628R
Human Ig lambda light chain Primary Antibody Unconjugated Host Species Rabbit Target Species Human Clonality Monoclonal Applications WB FCM IHC-P IHC-F IF(ICC) IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol