Learn More
Genetex Inc Rabbit anti-human IgA Monoclonal Antibody [MSVA-700R] HistoMAX™ (GTX04490) GeneTex - IHC-Optimized Antibody.
SDP
Catalog No. NC3668409
Encompass_Preferred
The Rabbit anti-human IgA antibody [MSVA-700R] HistoMAX™ (GTX04490) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human mucosa from duodenum tissue using GTX04490 Rabbit anti-human IgA antibody [MSVA-700R] HistoMAX. Human IgA positive plasma cells are abundant in the duodenum mucosa.
Catalog No. NC3668409 Supplier Genetex Inc Supplier No. GTX04490
The Rabbit anti-human IgA antibody [MSVA-700R] HistoMAX™ (GTX04490) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human mucosa from duodenum tissue using GTX04490 Rabbit anti-human IgA antibody [MSVA-700R] HistoMAX. Human IgA positive plasma cells are abundant in the duodenum mucosa.