Learn More
Bethyl Laboratories, Inc Human IgA ELISA Kit, 1 x 96 wells
Supplier:  Bethyl Laboratories, Inc E88102
Encompass_Preferred
CONTENTS:Human IgA Pre-Coated 96-well Strip Plate, 1 eachHuman IgA Standard, 750 ng/vial, 2 eachHuman IgA Detection Antibody, 12 ml10X Dilution Buffer B, 25 mlHRP Solution A, 12 mlTMB Substrate, 12 mlStop Solution, 12 ml20X Wash Buffer, 50 mlSealing Tape, 6 sheets
Catalog No. NC2881846