Learn More
Abcam Human IgA ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1962631X96TESTS
Encompass_Preferred
Human IgA ELISA kit is a single-wash 90-min SimpleStep ELISA used to quantification of human Immunoglobulin A (IgA) with a sensitivity of 0.25 ng/mL. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450nm readout; works on any plate reader - Different formats for different needs: 384-well for higher throughput - Broad sample compatibility – works with saliva, urine, plasma, cell culture supernatants, milk, and serum - Cited in over 10 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1777919