Learn More
Abcam Human IgA Matched Antibody Pair Kit, 5 x 96 Tests
Catalog No. 502587812
Encompass_Preferred
Human IgA Matched Antibody Pair Kit is a kit for ELISA development for the measurement of Human IgA by sandwich ELISA method.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-7812 Supplier Abcam Supplier No. AB2195365X96TESTS
Human IgA Matched Antibody Pair Kit is a kit for ELISA development for the measurement of Human IgA by sandwich ELISA method.
The product is subject to the following: Abcam Restricted Use Statement