Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IgA ProQuantum Immunoassay Kit

Catalog No. A457295
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A457295 5 x 96 Tests
A45729 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A457295 Supplier Invitrogen™ Supplier No. A457295
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human IgA, also known as human immunoglobulin A, is an antibody that is produced by the immune system in response to antigens. It is the most abundant immunoglobulin in mucosal secretions, such as saliva, tears, and breast milk, and plays a key role in defense against pathogens at mucosal surfaces. IgA works by binding to specific antigens and preventing them from entering the body's tissues. It can also activate complement and promote phagocytosis by immune cells. Human IgA can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious diseases.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01876, P01877
Assay Range 0.0256-2000 ng/mL
Assay Sensitivity <0.02 ng/mL
Conjugate FAM
Immunoassay Kit Format Proximity-based Immunoassay
Product Type ProQuantum
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) qPCR Instruments
Gene Alias A2m Marker; Ig alpha 1 Chain C Region; Ig alpha 2 Chain C Region; IgA1; IGHA1; IGHA2; Immunoglobulin A; Immunoglobulin Am1; Immunoglobulin Am2; immunoglobulin heavy constant alpha 1; Immunoglobulin Heavy Constant Alpha 2; immunoglobulin heavy constant alpha 2 (A2m marker)A2m Marker; Ig alpha 1 Chain C Region; Ig alpha 2 Chain C Region; IgA1; IGHA1; IGHA2; Immunoglobulin A; Immunoglobulin Am1; Immunoglobulin Am2; immunoglobulin heavy constant alpha 1; Immunoglobulin Heavy Constant Alpha 2; immunoglobulin heavy constant alpha 2 (A2m marker);
Gene ID (Entrez) 3493, 3494
Gene Symbol IGHA1, IGHA2
Kit Contents Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Label or Dye FAM™
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
Shipping Condition Dry Ice
Storage Requirements -20°C
Target Human IgA
Test Time 30 min
Total Assay Time 2 hr
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.