Learn More
Proteintech Group Inc human IgD Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc 675381IG150UL
Encompass_Preferred
The human IgD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human human IgD. This antibody recognizes human antigen. The human IgD antibody has been validated for the following applications: IF, IHC, WB,ELISA analysis.
Catalog No. 50-225-6518