Learn More
Meso Scale Discovery R-PLEX Human IgE Assay, 96-well SECTOR™ plates, analyte IgE, species reactivity human, assay format singleplex, LLOD 11.2 pg/mL, for research use only
SDP
Catalog No. NC2580685
Encompass
The MESO SCALE DISCOVERY R-PLEX Human IgE Assay is designed for the quantitative detection of human IgE through a singleplex electrochemiluminescence immunoassay. This assay offers exceptional sensitivity for biomarker discovery and development, and includes a complete set of reagents optimized for use with SECTOR plates and various MSD instruments. Intended for research use only, the assay is designed to support immunological and inflammatory research applications, making it a versatile tool for laboratories.
Catalog No. NC2580685 Supplier Meso Scale Discovery Supplier No. K1515AR2
The MESO SCALE DISCOVERY R-PLEX Human IgE Assay is designed for the quantitative detection of human IgE through a singleplex electrochemiluminescence immunoassay. This assay offers exceptional sensitivity for biomarker discovery and development, and includes a complete set of reagents optimized for use with SECTOR plates and various MSD instruments. Intended for research use only, the assay is designed to support immunological and inflammatory research applications, making it a versatile tool for laboratories.