Learn More
Invitrogen™ Human IgE ProQuantum Immunoassay Kit

Description
Specifications
Specifications
| Assay Range | 0.0077 to 3000 ng/mL |
| Assay Sensitivity | <10 pg/mL |
| Conjugate | FAM™ |
| Detection Method | Taqman™ Real-time PCR |
| Immunoassay Kit Format | Proximity-based Immunoassay |
| Product Type | ProQuantum |
| Sample Type | Plasma, Serum |
| Target Species | Human |
| For Use With (Application) | Protein Quantitation |
| For Use With (Equipment) | qPCR Instruments |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.