Learn More
Raybiotech Inc HUMAN IGF-1 ELISA
Supplier:  Raybiotech Inc ELHIGF11
Encompass_Preferred
RayBio Human IGF-1 ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 0.1 ng/ml - 30 ng/ml
Catalog No. 50-104-3909