Learn More
FUJIFILM BIOSCIENCES INC Human IGF-II (IGF-2), Recombinant Protein, Shenandoah Biotechnology, 50 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 1001450UG
Encompass_Preferred
Insulin-like growth factor II (IGF-II) is an important fetal growth hormone made by theca cells during gestation. IGF-II engages the IGF-I receptor (IGF1R) to mediate embryonic growth. IGF-II also binds the sink IGF-II receptor (IGF2R) leading to IGF-II degradation.
Catalog No. 50-990-667