Learn More
Gibco™ Human IGF-BP2 Recombinant Protein, PeproTech®
Description
350-06B-1MG will be provided as 2 x 500 μg (350-06B-500UG). Recombinant Human IGF-BP2 is a 31.5 kDa protein consisting of 289 amino acid residues including the IGF-BP domain and thyroglobulin type-I domain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis.
Specifications
Specifications
| Accession Number | P18065 |
| For Use With (Application) | Control, Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 3485 |
| Molecular Weight (g/mol) | 31.5 kDa |
| Name | Human IGF-BP2 |
| Purification Method | Purified |
| Quantity | 5 μg |
| Source | Insect cells |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.