Learn More
ACROBiosystems ACROBiosystems Human IGF-II Protein, Fc Tag
Supplier:  ACROBiosystems IG2H42601MG
Encompass_Preferred
Recombinant Protein;1MG;Human IGF-II, Fc Tag (IG2-H4260) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Glu 91 (Accession # P01344-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 34.4 kDa. The protein migrates as 36 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human IGFBP-3, His Tag (Cat. No. IG3-H5229) at 0.5 μg/mL (100 μL/well) can bind Human IGF-II, Fc Tag (Cat. No. IG2-H4260) with a linear range of 1-20 ng/mL (QC tested).
Catalog No. 50-210-6660