Learn More
Raybiotech Inc G-Series Human IGF Signaling Array 1 Kit.
Catalog No. 501950402
Encompass_Preferred
Semi-quantitative measurement of 10 human IGF family proteins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IGF-1, IGF-1 R, IGF-2, IGF-2 R, IGFBP-1, IGFBP-2, IGFBP-3, IGFBP-4, IGFBP-6, Insulin
Catalog No. 50-195-0402 Supplier Raybiotech Inc Supplier No. GSHIGF11
Semi-quantitative measurement of 10 human IGF family proteins. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IGF-1, IGF-1 R, IGF-2, IGF-2 R, IGFBP-1, IGFBP-2, IGFBP-3, IGFBP-4, IGFBP-6, Insulin