Learn More
Raybiotech Inc Quantibody Human Insulin-like Growth Factor Signaling Array 1 Kit.
Catalog No. 501950835
Encompass_Preferred
Detects 10 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: IGF-1, IGF-1 R, IGF-2, IGF-2 R, IGFBP-1, IGFBP-2, IGFBP-3, IGFBP-4, IGFBP-6, Insulin
Catalog No. 50-195-0835 Supplier Raybiotech Inc Supplier No. QAHIGF11
Detects 10 Human Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: IGF-1, IGF-1 R, IGF-2, IGF-2 R, IGFBP-1, IGFBP-2, IGFBP-3, IGFBP-4, IGFBP-6, Insulin