Learn More
Sino Biological Phospho-IGF1R (Tyr1131) / Insulin Receptor β (Tyr1146) Antibody, Rabbit MAb
Catalog No. 502992859
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic phosphopeptide corresponding to residues around Tyr1131 of the Human IGF1R or Tyr1146 of the Human Insulin Receptor β.
Catalog No. 50-299-2859 Supplier Sino Biological Supplier No. 111138R001920
This antibody was obtained from a rabbit immunized with a synthetic phosphopeptide corresponding to residues around Tyr1131 of the Human IGF1R or Tyr1146 of the Human Insulin Receptor β.