Learn More
BIOHIPPO Human IGF1R Microsample
SDP
Catalog No. 5000315622
Encompass
Human IGF1R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IGF1R in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 123 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IGF1R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IGF1R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IGF1R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15622 Supplier BIOHIPPO Supplier No. BHE15211730
Human IGF1R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IGF1R in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 123 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IGF1R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IGF1R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IGF1R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color