Learn More
Sino Biological Recombinant Human IGF1R Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10164H49H1B100
Encompass_Preferred
A DNA sequence encoding the human IGF1R (NP_000866.1) (Met1-Asn932) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1554