Learn More
Sino Biological Recombinant Human IGF1R Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10164H49HB20
Encompass_Preferred
A DNA sequence encoding the Human IGF1R (NP_000866.1)(Met1-Asn932) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-8194