Learn More
Sino Biological Recombinant Human IGF-II Protein (hFc & AVI Tag), Avi-tag Biotinylated, HPLC-verified
Catalog No. 502993462
Encompass_Preferred
A DNA sequence encoding the Human IGF2 (P01344-1)(Ala25-Glu91) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-3462 Supplier Sino Biological Supplier No. 13032H41HB20
A DNA sequence encoding the Human IGF2 (P01344-1)(Ala25-Glu91) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.