Learn More
Sino Biological Anti-IGF-II Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13032RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IGF-2 / IGF-II (rh IGF-2 / IGF-II; Catalog#13032-HNAY; P01344-1; Ala25-Glu91). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8870