Learn More
Raybiotech Inc Human IGF-2 R ELISA Kit
Supplier:  Raybiotech Inc ELHIGF2R1
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.27 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human Insulin-Like Growth Gactor type II Receptor.
Catalog No. 50-194-9378