Learn More
BIOHIPPO Human IGF2R Microsample
SDP
Catalog No. 5000313802
Encompass
Human IGF2R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IGF2R in Serum plasma and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 213 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IGF2R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IGF2R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IGF2R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13802 Supplier BIOHIPPO Supplier No. BHE15212112
Human IGF2R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IGF2R in Serum plasma and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 213 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IGF2R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IGF2R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IGF2R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color