Learn More
Sino Biological Recombinant Human Igf2r Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 14978H02H100
Encompass_Preferred
A DNA sequence encoding the Human Igf2r (P11717) (Met1-Asp1365) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-4112