Learn More
ACROBiosystems Human IGFBP-2 Protein His Tag
Supplier:  ACROBiosystems IG2H52H41MG
Encompass_Preferred
Human IGFBP-2 Protein His Tag (IG2-H52H4) is expressed from human 293 cells (HEK293). It contains AA Ala 36 - Gln325 (Accession # P18065).
Catalog No. 50-253-4638