Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Protein, Biotinylated Human IGFBP-7, His, Avitag™, 200 ug

Catalog No. 502719359
Encompass_Preferred

Insulin-like growth factor-binding protein 7 (IGFBP7) modulates the availability of insulin-like growth factors in tissues and regulates their binding to receptors. This human IGFBP-7 protein is expressed from HEK293 cells and contains an Ig-like C2-type domain, an IGFBP N-terminal domain, and a Kazal-like domain.

  • Expressed from human 293 cells
  • Contains a polyhistidine tag at the N-terminus followed by an Avi tag
  • Biotinylated using enzymatic labeling
  • Purity ≥95% as determined by SDS-PAGE and SEC-MALS
  • Calculated molecular weight of 29.8 kDa
  • Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose

Catalog No. 50-271-9359 Supplier ACROBiosystems Supplier No. IG7H82Q9200UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Insulin-like growth factor-binding protein 7 (IGFBP7) modulates the availability of insulin-like growth factors in tissues and regulates their binding to receptors. This human IGFBP-7 protein is expressed from HEK293 cells and contains an Ig-like C2-type domain, an IGFBP N-terminal domain, and a Kazal-like domain.

  • Expressed from human 293 cells
  • Contains a polyhistidine tag at the N-terminus followed by an Avi tag
  • Biotinylated using enzymatic labeling
  • Purity ≥95% as determined by SDS-PAGE and SEC-MALS
  • Calculated molecular weight of 29.8 kDa
  • Lyophilized from 0.22 μm filtered solution in PBS, pH 7.4 with trehalose

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.