Learn More
ACROBiosystems Biotinylated Human IGFBP-7 Protein, Fc,Avitag™ (MALS verified)
Catalog No. 502719361
Encompass_Preferred
Insulin-like growth factor-binding protein 7 (IGFBP7) is also known as IGFBP-rP1, MAC25 protein, PGI2-stimulating factor, prostacyclin-stimulating factor and tumor-derived adhesion factor, which contains one Ig-like C2-type (immunoglobulin-like) domain, one IGFBP N-terminal domain and one Kazal-like domain. The major function of IGFBP7 is the regulation of availability of insulin-like growth factors (IGFs) in tissue as well as in modulating IGF binding to its receptors. IGFBP7 binds to IGF with high affinity except for IGF-I and IGF-II.IGFBP7 also stimulates cell adhesion. Furthermore, IGFBP7 is implicated in some cancers.
Catalog No. 50-271-9361 Supplier ACROBiosystems Supplier No. IG7H82F9200UG
Insulin-like growth factor-binding protein 7 (IGFBP7) is also known as IGFBP-rP1, MAC25 protein, PGI2-stimulating factor, prostacyclin-stimulating factor and tumor-derived adhesion factor, which contains one Ig-like C2-type (immunoglobulin-like) domain, one IGFBP N-terminal domain and one Kazal-like domain. The major function of IGFBP7 is the regulation of availability of insulin-like growth factors (IGFs) in tissue as well as in modulating IGF binding to its receptors. IGFBP7 binds to IGF with high affinity except for IGF-I and IGF-II.IGFBP7 also stimulates cell adhesion. Furthermore, IGFBP7 is implicated in some cancers.