Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IGFBP-rp1/IGFBP-7 ELISA Kit

Catalog No. EH252RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH252RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH252RBX10 Supplier Invitrogen™ Supplier No. EH252RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human IGFBP-rp1/IGFBP-7 quantitates human IGFBP-rp1/IGFBP-7 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IGFBP-rp1/IGFBP-7.

This gene encodes a member of the insulin-like growth factor (IGF)-binding protein (IGFBP) family. IGFBPs bind IGFs with high affinity, and regulate IGF availability in body fluids and tissues and modulate IGF binding to its receptors. This protein binds IGF-I and IGF-II with relatively low affinity, and belongs to a subfamily of low-affinity IGFBPs. It also stimulates prostacyclin production and cell adhesion. Alternatively spliced transcript variants encoding different isoforms have been described for this gene, and one variant has been associated with retinal arterial macroaneurysm.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q16270
Assay Range 0.065-50 ng/mL
Assay Sensitivity 0.065 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AGM; angiomodulin; FSTL2; IBP7; IBP-7; IGF-binding protein 7; IGFBP; IGFBP rP1; Igfbp7; IGFBP-7; IGFBP-7v; IGFBPRP1; IGFBP-rP1; Insulin like growth factor binding protein; insulin like growth factor binding protein 7; insulin-like growth factor binding protein 7; insulin-like growth factor-binding protein 7; Mac25; MAC25 protein; PGI2-stimulating factor; prostacyclin-stimulating factor; PSF; RAMSVPS; TAF; Tumor-derived adhesion factorAGM; angiomodulin; FSTL2; IBP7; IBP-7; IGF-binding protein 7; IGFBP; IGFBP rP1; Igfbp7; IGFBP-7; IGFBP-7v; IGFBPRP1; IGFBP-rP1; Insulin like growth factor binding protein; insulin like growth factor binding protein 7; insulin-like growth factor binding protein 7; insulin-like growth factor-binding protein 7; Mac25; MAC25 protein; PGI2-stimulating factor; prostacyclin-stimulating factor; PSF; RAMSVPS; TAF; Tumor-derived adhesion factor;
Gene ID (Entrez) 3490
Gene Symbol IGFBP7
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target IGFBP7
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.