Learn More
Sino Biological Anti-IGFBP4 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10967MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IGFBP4 (rh IGFBP4; Catalog#10967-H08H; NP_001543.2; Met 1-Glu 258). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-5018