Learn More
Sino Biological Human IGFBP7 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502371727
Encompass_Preferred
Each vial contains 16 ng of recombinant Human IGFBP7. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 1000 pg/mL is recommended.
Catalog No. 50-237-1727 Supplier Sino Biological Supplier No. SEK1310015
Each vial contains 16 ng of recombinant Human IGFBP7. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 1000 pg/mL is recommended.