Learn More
Sino Biological Anti-IGFBP7 Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 13100MM01H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IGFBP7 (rh IGFBP7; Catalog#13100-H08H; Q16270; Met 1-Leu 282). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-2246