Learn More
Sino Biological Recombinant Anti-IGFBP7 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 13100R065100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IGFBP7 (rh IGFBP7; Catalog#13100-H08H; Q16270; Met1-Leu282).
Catalog No. 50-241-1279