Learn More
Sino Biological Anti-IGFBP7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13100RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IGFBP7 (rh IGFBP7; Catalog#13100-H08H; Q16270; Met1-Leu282). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1300