Learn More
Sino Biological Human IGFBP7 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 13100H56HB1
Encompass_Preferred
A DNA sequence encoding the Human IGFBP7 (Q16270) (Asp30-Leu282) was expressed with a N-terminal polyhistidine tag followed by an AVI tag.The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-1352