Learn More
Sino Biological Human IGFBP7 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 13100H41HB50
Encompass_Preferred
A DNA sequence encoding the human IGFBP7 (Q16270) (Met 1-Leu282) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-1786