Learn More
Rockland Immunochemicals HUMAN IgG whole molecule Biotin conjugated - 009-0602
SDP
Supplier:  Rockland Immunochemicals 0090602
Encompass
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The whole IgG molecule possesses both the F(c) region, recognized by high-affinity Fc receptor proteins, as well as the F(ab) region possessing the epitope-recognition site. Both heavy and light chains of the antibody molecule are present. This Human IgG whole molecule is conjugated to biotin (Vitamin H), a small biomolecule that has a large affinity for avidin and streptavidin.
Catalog No. NC0664425