Learn More
ACROBiosystems Human IgG control 100ug
Catalog No. 502535157
Encompass_Preferred
Human IgG1 (L234F L235E P331S) Kappa Isotype Control (mAb) is a chimeric monoclonal antibody recombinantly expressed from human 293 cells (HEK293) which combines the variable region of a mouse monoclonal antibody with human IgG1 constant domain. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP.
Catalog No. 50-253-5157 Supplier ACROBiosystems Supplier No. DNPC11P1100UG
Human IgG1 (L234F L235E P331S) Kappa Isotype Control (mAb) is a chimeric monoclonal antibody recombinantly expressed from human 293 cells (HEK293) which combines the variable region of a mouse monoclonal antibody with human IgG1 constant domain. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP.