Learn More
ACROBiosystems Human IgG1 (N297A) Kappa Isotype Control (mAb, carrier free, MALS verified)
Catalog No. 502331963
Encompass_Preferred
Human IgG1 (N297A) Kappa Isotype Control (mAb) is a chimeric monoclonal antibody recombinantly expressed from human 293 cells (HEK293), which combines the variable region of a mouse monoclonal antibody with human IgG1 constant domain. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP.
Catalog No. 50-233-1963 Supplier ACROBiosystems Supplier No. DNPMB27325MG
Human IgG1 (N297A) Kappa Isotype Control (mAb) is a chimeric monoclonal antibody recombinantly expressed from human 293 cells (HEK293), which combines the variable region of a mouse monoclonal antibody with human IgG1 constant domain. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP.