Learn More
Abcam Human IgG ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1952151X96TESTS
Encompass_Preferred
Human IgG ELISA kit is a single-wash, 90-minute SimpleStep ELISA assay designed for the quantification of human Immunoglobulin G (IgG) with a sensitivity of 0.02 ng/mL. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450nm readout; works on any plate reader - High-throughput options – available in 96-well and 384-well formats, supporting large-scale sample processing. - Broad sample compatibility – works with serum, plasma, cell culture supernatants, and other biological fluids - Cited in over 20 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0986215