Learn More
Rockland Immunochemicals Human IgG F(ab')2 2 mg
SDP
Supplier:  Rockland Immunochemicals 0090104
Encompass
Human IgG F(ab')2 purified protein is a proteolytic fragment of immunoglobulin G (IgG) obtained by limited digestion with the enzyme pepsin under controlled conditions of temperature time and pH. Human IgG F(ab')2 molecules lack the Fc portion of Human IgG and therefore receptors that bind Human IgG Fc will not bind Human IgG F(ab')2 molecules.
Catalog No. 50-194-3754