Learn More
Rockland Immunochemicals HUMAN IgG Fab fragment - 009-0105
SDP
Catalog No. 501057327
Encompass_Preferred
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The F(ab) fragment is the portion of the antibody that binds to the antigen target. The immunoglobulin Fab also possesses one constant and one variable region of both the heavy and light chain.
Catalog No. 50-105-7327 Supplier Rockland Immunochemicals Supplier No. 0090105
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The F(ab) fragment is the portion of the antibody that binds to the antigen target. The immunoglobulin Fab also possesses one constant and one variable region of both the heavy and light chain.