Learn More
Rockland Immunochemicals Human IgG (H&L) Secondary Antibody Peroxidase Conjugated Pre-Adsorbed 1 mg
SDP
Supplier:  Rockland Immunochemicals 609103123
Encompass
Anti-Human IgG (H&L) Peroxidase generated in goat detects human Immunoglobulin G (IgG) both heavy and light chains of the antibody molecule are present. It is a protein complex composed of four peptide chains — two identical heavy chains and two identical light chains arranged in a Y-shape typical of antibody monomers. Each IgG has two antigen binding sites. Representing approximately 75% of serum immunoglobulins in humans IgG is the most abundant antibody isotype found in the circulation. IgG molecules are synthesized and secreted by plasma B cells. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.
Catalog No. 50-194-3885