Learn More
Invitrogen™ Human IgG Isotype Control
Description
Specificity was determined by ELISA using a panel of antibodies specific to human IgG, IgA, IgM, Kappa and Lambda. Concentration provided: 1 mg/mL. Antibody value assigned is based on the extinction coefficient of IgG: Optical Density at 280 nm of 1.4 equals 1 mg antibody.
Specifications
Specifications
| Antigen | Human IgG |
| Applications | Control, Flow Cytometry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene Alias | Immunoglobulin G; ImmunoglobulinG |
| Host Species | Human |
| Immunogen | Gamma Immunoglobin |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.