Learn More
Invitrogen™ Human IgG Isotype Control
Description
Human IgG is purified from pooled normal human serum. The protein concentration is 5.0 mg/mL based on an E (1%) = 14.0 at 280 nm.
Specifications
Specifications
| Antigen | Human IgG |
| Applications | Control, Flow Cytometry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene Alias | Immunoglobulin G; ImmunoglobulinG |
| Host Species | Human |
| Immunogen | Gamma Immunoglobin |
| Purification Method | Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.